Learn More
ELITechGroup Aerospray Hematology Stat Reagent A, Basofix™ Rinse, 500mL
SDP
Catalog No. NC9137906
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Orange; 6 +/- 0.2 pH; Hematology Reagent A Basofix Rinse; B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light
Catalog No. NC9137906 Supplier ELITechGroup Supplier No. SS049A
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Orange; 6 +/- 0.2 pH; Hematology Reagent A Basofix Rinse; B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light