Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Basolite™ F300

Catalog No. 69087210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-087-210G 10 g
69-087-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-087-210G Supplier Sigma Aldrich Supplier No. 69087210G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Fe-BTC based MOF can be used in greener applications such as energy storage through lithium ion batteries. It can also be used in the supercritical adsorption of CO2.Basolite is a registered trademark of BASF SE

Specifications

CAS 1195763-37-1
Molecular Formula C9H3FeO6
Linear Formula C9H3FeO6
Quantity 10 g
MDL Number MFCD11656051
Synonym Fe-BTC; Iron 1,3,5-benzenetricarboxylate
Molecular Weight (g/mol) 262.96
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.