Learn More
Aviva Systems Biology Corp BASP1 Antibody - C-terminal region (ARP59932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BASP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BASP1. Alternate names: CAP-23, CAP23, MGC8555, NAP-22, NAP22ApplicationWB. Species reactivity: Human, Rabbit, Rat, Yeast
Catalog No. 50-286-881