Learn More
Enzo Life Sciences Bassoon monoclonal antibody (SAP7F407) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6660200
Encompass_Preferred
Clone: SAP7F407. Isotype: IgG2aκ. Immunogen: Recombinant rat Bassoon. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: ICC, IF, IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3192