Learn More
Enzo Life Sciences Bassoon monoclonal antibody (SAP7F407) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPS003F
Encompass_Preferred
Clone: SAP7F407. Host: Mouse. Isotype: IgG2aκ. Immunogen: Recombinant rat Bassoon. UniProt: O88778. Source: Purified from ascites. Species Reactivity: Mouse, Rat. Applications: ICC, IF, IHC, IP, WB. Recommended dilutions: Immunocytochemistry (1:400). Immunoprecipitation (2.5µg). Western Blot (1:500 - 1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-832