Learn More
Bioworld Glufosinate Ammonium, 250 mg
Catalog No. 50255088
Encompass_Preferred
Specification:Synonym: Glufosinate ammonium, Phosphinothricin (PPT)Non-selective herbicide for plant transformation studies using nucleotide sequence of the phosphinothricin N-acetyltransferase gene from Streptomyces viridochromogenes, "Bar gene"Acts by inhibiting glutamine synthetase and thus blocks all nitrogen assimilation into the plantBasta is the registered trademark of Bayer CropScience
Catalog No. 50-255-088 Supplier Bioworld Supplier No. 306320074
Specification:Synonym: Glufosinate ammonium, Phosphinothricin (PPT)Non-selective herbicide for plant transformation studies using nucleotide sequence of the phosphinothricin N-acetyltransferase gene from Streptomyces viridochromogenes, "Bar gene"Acts by inhibiting glutamine synthetase and thus blocks all nitrogen assimilation into the plantBasta is the registered trademark of Bayer CropScience