Learn More
Kingfisher Biotech Inc Black Flying Fox Bat CCL3 ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891925
Encompass_Preferred
The Black Flying Fox Bat CCL3 (MIP-1 alpha) ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1925 Supplier Kingfisher Biotech Inc Supplier No. SPOT2322BT004
The Black Flying Fox Bat CCL3 (MIP-1 alpha) ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA