Learn More
Kingfisher Biotech Inc Bovine CXCL9 (MIG) ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY2067BT004
Encompass_Preferred
The Bovine CXCL9 (MIG) ELISA contains capture antibody, protein standard, and detection antibody for development of a Bovine CXCL9 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine CXCL9 (MIG) ELISA can also be used to measure Bison, Wild Yak, and Zebu CXCL9 (MIG). Alternate names include CXCL9, CMK, Humig, MIG, SCYB9, crg-10, C-X-C motif chemokine ligand 9. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5672