Learn More
Aviva Systems Biology Corp BAT1 Antibody - C-terminal region (ARP36372_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36372_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BAT1. Alternate names: BAT1, UAP56, D6S81EApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56131