Learn More
Aviva Systems Biology Corp BAT3 Antibody - C-terminal region (ARP61643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAT3. It was validated on Western Blot. . Alternate names: BAG-6, BAG6, D6S52E, G3, BAT3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-431