Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Bata Viking Galoshes and Overboots

Catalog No. 17986369 Shop All Onguard Industries LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
2X-Large
For Use With (Equipment):
Size 14 to 15 shoes
Height (English):
10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Equipment) Height (English)
17-986-369 2X-Large Size 14 to 15 shoes 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-986-369 Supplier Dunlop™ Supplier No. 860202X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Closure Type Plastic snaps
Color Black
For Use With (Application) Used in construction, farming, utilities and food processing
For Use With (Equipment) Size 14 to 15 shoes
Height (English) 10 in.
Lug Type Kick-off
Material Flexothane, PVC
Size 2X-Large
Sole Features 4-way cleated, slip-resistant outsole
Toe Style Plain
Waterproof Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.