Learn More
Medchemexpress LLC Batabulin | 195533-53-0 | MFCD22380604 | 99.9% | C13H7F6NO3S | 10MG
SDP
Catalog No. 5000258580
Encompass_Preferred
Batabulin is a covalent microtubule-targeting antitumor reagent for laboratory research. It selectively binds a subset of β-tubulin isotypes, disrupts microtubule polymerization, alters cell morphology, and can induce cell-cycle arrest and apoptosis. Supplied for research use only. Molecular formula C13H7F6NO3S, MW 371.26, CAS 195533-53-0.
Catalog No. 50-002-58580 Supplier Medchemexpress LLC Supplier No. HY1356310MG
Batabulin is a covalent microtubule-targeting antitumor reagent for laboratory research. It selectively binds a subset of β-tubulin isotypes, disrupts microtubule polymerization, alters cell morphology, and can induce cell-cycle arrest and apoptosis. Supplied for research use only. Molecular formula C13H7F6NO3S, MW 371.26, CAS 195533-53-0.