Learn More
Medchemexpress LLC HY-13563 5mg Medchemexpress, Batabulin CAS:195533-53-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135635MG
Encompass_Preferred
Medchemexpress, HY-13563 5mg Batabulin CAS:195533-53-0 Batabulin (T138067) is an antitumor agent, which binds covalently and selectively to a subset of the β-tubulin isotypes, thereby disrupting microtubule polymerization. Batabulin affects cell morphology and leads to cell-cycle arrest ultimately induces apoptotic cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0346