Learn More
Medchemexpress LLC Batabulin sodium | 195533-98-3 | MFCD22380600 | 99.8% | 393.24 g/mol | C13H6F6NNaO3S | 5 MG
SDP
Catalog No. 5000222722
Encompass_Preferred
Batabulin sodium is a sodium salt antitumor research compound that covalently and selectively binds specific β-tubulin isotypes, disrupting microtubule polymerization, inducing G2/M cell-cycle arrest, and triggering apoptotic cell death. Supplied as an off-white to light-yellow solid for in vitro and in vivo research applications.
  • Covalently binds to a subset of β-tubulin isotypes.
  • Disrupts microtubule polymerization and induces G2/M arrest.
  • Induces apoptosis in cell-based assays.
  • High reported purity (~99.8%).
  • Soluble in DMSO; use freshly opened solvent and ultrasonic assistance if needed.
  • Store powder at 4°C; in solution store at -80°C for long-term stability.
Catalog No. 50-002-22722 Supplier Medchemexpress LLC Supplier No. HY13563A5MG
Batabulin sodium is a sodium salt antitumor research compound that covalently and selectively binds specific β-tubulin isotypes, disrupting microtubule polymerization, inducing G2/M cell-cycle arrest, and triggering apoptotic cell death. Supplied as an off-white to light-yellow solid for in vitro and in vivo research applications.
  • Covalently binds to a subset of β-tubulin isotypes.
  • Disrupts microtubule polymerization and induces G2/M arrest.
  • Induces apoptosis in cell-based assays.
  • High reported purity (~99.8%).
  • Soluble in DMSO; use freshly opened solvent and ultrasonic assistance if needed.
  • Store powder at 4°C; in solution store at -80°C for long-term stability.