Learn More
Medchemexpress LLC Batabulin sodium | 195533-98-3 | MFCD22380600 | 99.8% | C13H6F6NNaO3S | 10MG
SDP
Catalog No. 5000258193
Encompass_Preferred
Batabulin sodium is the sodium salt of batabulin, a tubulin-targeting antitumor research compound that covalently binds a subset of β-tubulin isotypes and disrupts microtubule dynamics, inducing apoptosis in cell models. It is supplied as a high-purity reagent for biochemical and cellular studies of microtubules, tubulin covalent modification, and related pharmacology.
  • Covalently binds a subset of β-tubulin isotypes and disrupts microtubule dynamics.
  • Used to study apoptosis, cell division, and tubulin-targeting pharmacology.
  • Supplied as a high-purity research reagent (99.8%).
  • Provided in multiple pack sizes and concentration formats for lab use.
  • Sodium salt form suitable for biochemical and cell-based assays.
Catalog No. 50-002-58193 Supplier Medchemexpress LLC Supplier No. HY13563A10MG
Batabulin sodium is the sodium salt of batabulin, a tubulin-targeting antitumor research compound that covalently binds a subset of β-tubulin isotypes and disrupts microtubule dynamics, inducing apoptosis in cell models. It is supplied as a high-purity reagent for biochemical and cellular studies of microtubules, tubulin covalent modification, and related pharmacology.
  • Covalently binds a subset of β-tubulin isotypes and disrupts microtubule dynamics.
  • Used to study apoptosis, cell division, and tubulin-targeting pharmacology.
  • Supplied as a high-purity research reagent (99.8%).
  • Provided in multiple pack sizes and concentration formats for lab use.
  • Sodium salt form suitable for biochemical and cell-based assays.