Learn More
Aviva Systems Biology Corp BATF Antibody - middle region (ARP32772_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BATF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BATF. Alternate names: B-ATF, BATF1, SFA-2, SFA2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54754