Learn More
Aviva Systems Biology Corp BATF2 Antibody - middle region (ARP47712_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47712_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BATF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BATF2. Alternate names: MGC20410, SARIApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-53526