Learn More
ABclonal Technology BATF3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A149061000UL
Encompass_Preferred
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription.
Catalog No. 50-001-05646