Learn More
ABclonal Technology BATF3 Rabbit pAb
SDP
Catalog No. 5000105217
Encompass_Preferred
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription.
Catalog No. 50-001-05217 Supplier ABclonal Technology Supplier No. A14906500UL
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription.