Learn More
Cole-Parmer Techne FBATH18 Unheated Water Bath, Stainless Steel, 18L
Supplier:  Cole-Parmer 1611201
Encompass
Designed to be used with a clip-on Tempette or Tempunit thermoregulator (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
Catalog No. 50-196-0476