Learn More
Cole-Parmer Essentials Hollow PolyPropylene (PP) Ball Bath Covers, 10 mm; 2500/Pk
Catalog No. NC2090276 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.
This item is not returnable. View return policy
Catalog No. NC2090276 Supplier Cole-Parmer Supplier No. 0682100
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.