Learn More
MSC Detco Industries Insect Repellent, Repell More II, Aerosol, 1 ea
Catalog No. 502815428
Encompass
Repell More II is an insect repellent formulated as a flammable aerosol, designed for general use. It contains active ingredients including Deet, Mgk 264, and Di-n-propyl Isocinchomeronate.
  • Contains active ingredients Deet, Mgk 264, and Di-n-propyl Isocinchomeronate
  • Provided as a pressurized aerosol liquid/gas
  • Designed for general insect repellant applications
  • Flammable aerosol, contains liquefied gases
Catalog No. 50-281-5428 Supplier MSC Supplier No. 33687781
Repell More II is an insect repellent formulated as a flammable aerosol, designed for general use. It contains active ingredients including Deet, Mgk 264, and Di-n-propyl Isocinchomeronate.
  • Contains active ingredients Deet, Mgk 264, and Di-n-propyl Isocinchomeronate
  • Provided as a pressurized aerosol liquid/gas
  • Designed for general insect repellant applications
  • Flammable aerosol, contains liquefied gases