Learn More
Polyscience polycool EG -25
SDP
Catalog No. NC9964619
Encompass_Preferred
PolyScience™ PolyScience° polycool EG-25 -25 to 100°C Temperature Range1 gallon (3.8L) BottleFeatures:• A common circulator antifreeze fluid.• Lowers the freezing point of water to allow circulation at below freezing temperatures.High toxicity.• Recommend mixing with distilled water for broadest temperature range
Catalog No. NC9964619 Supplier Polyscience Supplier No. 060340
PolyScience™ PolyScience° polycool EG-25 -25 to 100°C Temperature Range1 gallon (3.8L) BottleFeatures:• A common circulator antifreeze fluid.• Lowers the freezing point of water to allow circulation at below freezing temperatures.High toxicity.• Recommend mixing with distilled water for broadest temperature range