Learn More
Polyscience polycool HC -50
SDP
Catalog No. NC0120248
Encompass_Preferred
PolyScience™ PolyScience° polycool HC -50 Temperature Range: -50 to 100°C)1 gallon (3.8 L) BottleFeatures:• Excellent low-temperature performance without toxicity or risk to environment, equipment or personnel.• Provides low-temp properties of synthetic organic and silicone fluids with thermal properties of water-based glycols.
Catalog No. NC0120248 Supplier Polyscience Supplier No. 060330
PolyScience™ PolyScience° polycool HC -50 Temperature Range: -50 to 100°C)1 gallon (3.8 L) BottleFeatures:• Excellent low-temperature performance without toxicity or risk to environment, equipment or personnel.• Provides low-temp properties of synthetic organic and silicone fluids with thermal properties of water-based glycols.