Learn More
Cole-Parmer Fluke Calibration 6102 Portable Calibration Bath; 35 to 200C
Catalog No. 50992495 Shop All Cole-Parmer Products
Encompass
These calibration baths are designed with the stability and accuracy of a fluid bath and the compact size and weight of a portable calibrator. Smaller and lighter than most dry-well calibrators, these calibration baths are easy to carry and are spill-proof. The heating, cooling, and stabilization times allow you to take readings within 45 minutes. Bath is ideal for glass and bimetal thermometers. There’s no need to make custom well inserts — the 2-½" diameter stirred tank holds just about any size probe. All calibration baths have built-in RS-232 interface and include software for Windows 3.x/95. The basic data collection software allows you to change the temperature, set points, and safety cutouts on your bath. Available for sale in the United States and Canada only.
Catalog No. 50-992-495 Supplier Cole-Parmer Supplier No. 9091200
Add to Cart
Add to Cart
Request a Quote
These calibration baths are designed with the stability and accuracy of a fluid bath and the compact size and weight of a portable calibrator. Smaller and lighter than most dry-well calibrators, these calibration baths are easy to carry and are spill-proof. The heating, cooling, and stabilization times allow you to take readings within 45 minutes. Bath is ideal for glass and bimetal thermometers. There’s no need to make custom well inserts — the 2-½" diameter stirred tank holds just about any size probe. All calibration baths have built-in RS-232 interface and include software for Windows 3.x/95. The basic data collection software allows you to change the temperature, set points, and safety cutouts on your bath. Available for sale in the United States and Canada only.