Learn More
Julabo USA Inc BATH TANK BT19 19L
SDP
Catalog No. NC2329048
Encompass_Preferred
19 L Transparent bath tank up to +100°C for heating immersion circulators CORIO C and CD. Inner dimensions (WxL/D): 11.8 x 19.6 / 5.9″; Outer dimensions (WxL/D): 15 x 22.8 x 6.3″.
Catalog No. NC2329048 Supplier Julabo USA Inc Supplier No. 9901319
19 L Transparent bath tank up to +100°C for heating immersion circulators CORIO C and CD. Inner dimensions (WxL/D): 11.8 x 19.6 / 5.9″; Outer dimensions (WxL/D): 15 x 22.8 x 6.3″.