Learn More
Cole-Parmer Cole-Parmer Bath Thermal Transfer Fluid, Ethylene Glycol / DI Water Blend – 50:50 Mix; 4L Poly Bottle
Catalog No. NC3788326 Shop All Cole-Parmer Products
Encompass_Preferred
Use bath fluids to maintain desired bath temperatures outside the range of water.Lower freezing point than water.50/50 Ethylene Glycol/WaterThermal transfer fluid for -25C to 100CFor use with circulating baths and chillers
Catalog No. NC3788326 Supplier Cole-Parmer Supplier No. 7893017
Use bath fluids to maintain desired bath temperatures outside the range of water.Lower freezing point than water.50/50 Ethylene Glycol/WaterThermal transfer fluid for -25C to 100CFor use with circulating baths and chillers