Learn More
Decon Labs BathBrite
GREENER_CHOICE
Catalog No. 04355142 Shop All Decon Labs Products
Change view
Click to view available options
Size:
8 oz.
Catalog No. Size
04-355-142 8 oz.
1 options
This item is not returnable. View return policy
Catalog No. 04-355-142 Supplier Decon Labs Supplier No. 3408
  • PRODUCT PROMOTION
    $744.00
    $342.89 / Case of 6
    Save $401.11
    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Maintains clean, fresh water in laboratory water baths and incubators with minimal ongoing effort.
  • Designed specifically for use in water baths and incubators to support routine laboratory maintenance
  • Requires only 10 to 15 drops per application for effective performance
  • Helps prevent water contamination, extending the interval between complete water replacements
  • Simple to use additive that supports consistent operating conditions in lab equipment
Specifications
Product Type Cleaner
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Container Type Squeeze Bottle
Quantity 6/Cs.
Size 8 oz.