Learn More
Medchemexpress LLC Bathocuproine disulfonate disodium | 52698-84-7 | MFCD00011679 | 99.5% | 564.54 g·mol⁻¹ | C26H18N2Na2O6S2 | 1 G
SDP
Catalog No. 5000300281
Encompass_Preferred
Bathocuproine disulfonate disodium is a water-soluble sulfonated derivative of bathocuproine used as a selective chelator and colorimetric reagent for copper ions in biochemical and analytical assays. It is intended for laboratory research applications requiring sensitive detection or sequestration of copper.
  • Water-soluble copper chelator suitable for colorimetric and spectrophotometric assays.
  • High reported purity (99.53%), reducing potential assay interference.
  • Soluble and stable in aqueous solutions for typical laboratory use.
  • Molecular weight 564.54 g·mol⁻¹ and formula C26H18N2Na2O6S2 for accurate stoichiometry.
  • Available in small pack sizes convenient for research workflows.
Catalog No. 50-003-00281 Supplier Medchemexpress LLC Supplier No. HYW0349531G
Bathocuproine disulfonate disodium is a water-soluble sulfonated derivative of bathocuproine used as a selective chelator and colorimetric reagent for copper ions in biochemical and analytical assays. It is intended for laboratory research applications requiring sensitive detection or sequestration of copper.
  • Water-soluble copper chelator suitable for colorimetric and spectrophotometric assays.
  • High reported purity (99.53%), reducing potential assay interference.
  • Soluble and stable in aqueous solutions for typical laboratory use.
  • Molecular weight 564.54 g·mol⁻¹ and formula C26H18N2Na2O6S2 for accurate stoichiometry.
  • Available in small pack sizes convenient for research workflows.