Learn More
Medchemexpress LLC Bathocuproine disulfonate disodium | 52698-84-7 | MFCD00011679 | 99.5% | 564.54 g·mol⁻¹ | C26H18N2Na2O6S2 | 100 MG
SDP
Catalog No. 5000299421
Encompass_Preferred
Bathocuproine disulfonate disodium is a water-soluble chelator used as a highly sensitive colorimetric reagent for detection and quantification of copper ions in biochemical, analytical, and environmental assays. It forms a colored complex with copper that enables spectrophotometric measurements and is supplied as a high-purity solid for laboratory use.
  • Highly sensitive colorimetric detection of copper ions.
  • Water-soluble disodium salt suitable for aqueous assays.
  • High purity for reproducible analytical results.
  • Available in multiple pack sizes for lab-scale use.
  • Recommended storage protects stability: keep cool, protect from light, store under inert gas.
Catalog No. 50-002-99421 Supplier Medchemexpress LLC Supplier No. HYW034953100MG
Bathocuproine disulfonate disodium is a water-soluble chelator used as a highly sensitive colorimetric reagent for detection and quantification of copper ions in biochemical, analytical, and environmental assays. It forms a colored complex with copper that enables spectrophotometric measurements and is supplied as a high-purity solid for laboratory use.
  • Highly sensitive colorimetric detection of copper ions.
  • Water-soluble disodium salt suitable for aqueous assays.
  • High purity for reproducible analytical results.
  • Available in multiple pack sizes for lab-scale use.
  • Recommended storage protects stability: keep cool, protect from light, store under inert gas.