Learn More
Medchemexpress LLC Bathocuproine disulfonate disodium | 52698-84-7 | MFCD00011679 | 99.5% | 564.54 g·mol⁻¹ | C26H18N2Na2O6S2 | 250 MG
SDP
Catalog No. 5000303468
Encompass_Preferred
Bathocuproine disulfonate disodium is the disodium salt of bathocuproine disulfonic acid, a water-soluble copper(I) chelator used as a colorimetric reagent to detect and quantify copper ions in biochemical and analytical assays. It is supplied as a solid reagent for research applications.
  • High purity: 99.5%.
  • Water-soluble disodium salt for aqueous assays.
  • Specific chelation and colorimetric response to copper(I) ions.
  • Useful for quantitative copper determination in biochemical samples.
  • Available in multiple sizes suitable for small-scale research.
  • Stable when stored protected from light at low temperature.
Catalog No. 50-003-03468 Supplier Medchemexpress LLC Supplier No. 250MGHYW034953
Bathocuproine disulfonate disodium is the disodium salt of bathocuproine disulfonic acid, a water-soluble copper(I) chelator used as a colorimetric reagent to detect and quantify copper ions in biochemical and analytical assays. It is supplied as a solid reagent for research applications.
  • High purity: 99.5%.
  • Water-soluble disodium salt for aqueous assays.
  • Specific chelation and colorimetric response to copper(I) ions.
  • Useful for quantitative copper determination in biochemical samples.
  • Available in multiple sizes suitable for small-scale research.
  • Stable when stored protected from light at low temperature.