Learn More
Medchemexpress LLC Bathocuproine disulfonate disodium | 52698-84-7 | MFCD00011679 | 99.5% | 564.54 g·mol⁻¹ | C26H18N2Na2O6S2 | 500 MG
SDP
Catalog No. 5000300174
Encompass_Preferred
Bathocuproine disulfonate disodium (BCS) is a water-soluble, sulfonated phenanthroline derivative that selectively chelates Cu(I) and is used as a colorimetric and spectrophotometric reagent to detect and quantify copper ions in biochemical and analytical assays. Supplied as a solid with high stated purity, it is intended for laboratory analytical applications.
  • Selective chelator for copper(I) ions.
  • Water-soluble sulfonated phenanthroline derivative.
  • High purity suitable for analytical assays.
  • Off-white to light brown solid form.
  • Store at 4°C, protect from light, and under nitrogen for stability.
Catalog No. 50-003-00174 Supplier Medchemexpress LLC Supplier No. HYW034953500MG
Bathocuproine disulfonate disodium (BCS) is a water-soluble, sulfonated phenanthroline derivative that selectively chelates Cu(I) and is used as a colorimetric and spectrophotometric reagent to detect and quantify copper ions in biochemical and analytical assays. Supplied as a solid with high stated purity, it is intended for laboratory analytical applications.
  • Selective chelator for copper(I) ions.
  • Water-soluble sulfonated phenanthroline derivative.
  • High purity suitable for analytical assays.
  • Off-white to light brown solid form.
  • Store at 4°C, protect from light, and under nitrogen for stability.