Learn More
Crescent Chemical Co Inc BATHOCUPROINE DISULFONIC ACID
SDP
Supplier:  Crescent Chemical Co Inc SE14470.02
Encompass_Preferred
Bathocuproine disulfonic acid-Na2-salt analytical grade Copper detection in polyacrylamide gels. Size - 1G Storage Conditions - +15 °C TO +30 °C Catalog Number - 14470.02 CAS 52698-84-7
Catalog No. 50-247-333