Learn More
Sigma Aldrich Fine Chemicals Biosciences Bathocuproinedisulfonic acid disodium salt for spectrophotometric det. of Cu, Fe | Purity: 7.2-9.1% dry basis (ICP) | Mol Wt: 564.54 | 52698-84-7 | 500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences B1125500MG
Encompass
Bathocuproinedisulfonic acid disodium salt for spectrophotometric det. of Cu, Fe | Purity: 7.2-9.1% dry basis (ICP) | Mol Wt: 564.54 | 52698-84-7 | 500MG
Catalog No. 50-186-7155