Learn More
Apexbio Technology LLC Bathophenanthroline 1662-01-7 1g
SDP
Catalog No. 5000090706
Encompass_Preferred
Bathophenanthroline (CAS 1662-01-7) is an organic small-molecule reagent designed for selective detection of ferrous ions (Fe ) via colorimetric chelation It forms stable colored complexes with Fe allowing quantification of iron concentrations in various matrices Bathophenanthroline exerts its analytical activity through specific chelation of Fe producing colorimetric changes measurable at defined wavelengths Based on these properties bathophenanthroline holds research potential in the quantitative assessment of iron in biological and environmental samples supporting applications in studies of iron homeostasis metabolic pathways oxidative stress assessment and clinical diagnostics of iron metabolism disorders
Catalog No. 50-000-90706 Supplier Apexbio Technology LLC Supplier No. C66661000
Bathophenanthroline (CAS 1662-01-7) is an organic small-molecule reagent designed for selective detection of ferrous ions (Fe ) via colorimetric chelation It forms stable colored complexes with Fe allowing quantification of iron concentrations in various matrices Bathophenanthroline exerts its analytical activity through specific chelation of Fe producing colorimetric changes measurable at defined wavelengths Based on these properties bathophenanthroline holds research potential in the quantitative assessment of iron in biological and environmental samples supporting applications in studies of iron homeostasis metabolic pathways oxidative stress assessment and clinical diagnostics of iron metabolism disorders