Learn More
Apexbio Technology LLC Bathophenanthroline 1662-01-7 500mg
SDP
Catalog No. 5000090131
Encompass_Preferred
Bathophenanthroline (CAS 1662-01-7) is an organic small-molecule reagent designed for selective detection of ferrous ions (Fe ) via colorimetric chelation It forms stable colored complexes with Fe allowing quantification of iron concentrations in various matrices Bathophenanthroline exerts its analytical activity through specific chelation of Fe producing colorimetric changes measurable at defined wavelengths Based on these properties bathophenanthroline holds research potential in the quantitative assessment of iron in biological and environmental samples supporting applications in studies of iron homeostasis metabolic pathways oxidative stress assessment and clinical diagnostics of iron metabolism disorders
Catalog No. 50-000-90131 Supplier Apexbio Technology LLC Supplier No. C6666500
Bathophenanthroline (CAS 1662-01-7) is an organic small-molecule reagent designed for selective detection of ferrous ions (Fe ) via colorimetric chelation It forms stable colored complexes with Fe allowing quantification of iron concentrations in various matrices Bathophenanthroline exerts its analytical activity through specific chelation of Fe producing colorimetric changes measurable at defined wavelengths Based on these properties bathophenanthroline holds research potential in the quantitative assessment of iron in biological and environmental samples supporting applications in studies of iron homeostasis metabolic pathways oxidative stress assessment and clinical diagnostics of iron metabolism disorders