Learn More
Medchemexpress LLC Bathophenanthroline | 1662-01-7 | 99.6% | 500 MG
SDP
Catalog No. 5000409670
Encompass_Preferred
Bathophenanthroline is a nitrogen-containing organic ligand that forms stable complexes with transition metal ions like Ru2+. It is used in proteomics for in-gel protein staining, where its negatively charged sulfonic acid groups electrostatically interact with positively charged amino acid residues, enhancing fluorescent detection signals.
  • Forms stable complexes with transition metal ions.
  • Used for in-gel protein staining in proteomics.
  • Enhances fluorescent detection signals.
  • Can be used for protein separation and detection in SDS-PAGE gels.
  • Suitable for other fluorescence studies.
  • Excitation/emission at 532 nm/610 nm.
  • Shows higher sensitivity than SYPRO Ruby for protein detection at near-neutral pH.
  • Reduces artificial post-translational modifications.
Catalog No. 50-004-09670 Supplier Medchemexpress LLC Supplier No. HYD0006500MG
Bathophenanthroline is a nitrogen-containing organic ligand that forms stable complexes with transition metal ions like Ru2+. It is used in proteomics for in-gel protein staining, where its negatively charged sulfonic acid groups electrostatically interact with positively charged amino acid residues, enhancing fluorescent detection signals.
  • Forms stable complexes with transition metal ions.
  • Used for in-gel protein staining in proteomics.
  • Enhances fluorescent detection signals.
  • Can be used for protein separation and detection in SDS-PAGE gels.
  • Suitable for other fluorescence studies.
  • Excitation/emission at 532 nm/610 nm.
  • Shows higher sensitivity than SYPRO Ruby for protein detection at near-neutral pH.
  • Reduces artificial post-translational modifications.