Learn More
Medchemexpress LLC HY-D0006 1g Medchemexpress, Bathophenanthroline CAS:1662-01-7 Purity:>98%
SDP
Catalog No. 501933129
Encompass_Preferred
Medchemexpress, HY-D0006 1g Bathophenanthroline CAS:1662-01-7 Bathophenanthroline is an agent used for the measurement of iron concentrations in aqueous , serum and urine samples by colorimetry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3129 Supplier Medchemexpress LLC Supplier No. HYD00061G
Medchemexpress, HY-D0006 1g Bathophenanthroline CAS:1662-01-7 Bathophenanthroline is an agent used for the measurement of iron concentrations in aqueous , serum and urine samples by colorimetry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.