Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bathophenanthroline (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. B26951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26951G Supplier TCI America Supplier No. B26951G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Specifications

Melting Point 221°C
Color White
Quantity 1 g
Formula Weight 332.41
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bathophenanthroline (purified by sublimation)
RTECSNumber SF8427000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.