Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bathophenanthrolinedisulfonic acid disodium salt hydrate

Catalog No. 1466175G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-661-75G 5 g
14-661-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-661-75G Supplier Sigma Aldrich Supplier No. 1466175G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bathophenanthrolinedisulfonic acid disodium salt hydrate

Specifications

CAS 52746-49-3
Melting Point 300°C
Molecular Formula C24H14N2Na2O6S2 -+ 3H2O
Linear Formula C24H14N2Na2O6S2 -+ 3H2O
Quantity 5 g
Synonym Bathophenanthrolinedisulfonic acid disodium salt trihydrate; 4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate; Disodium bathophenanthrolinedisulfonate trihydrate
Molecular Weight (g/mol) 590.53
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.