Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bathophenanthrolinedisulfonic acid disodium salt trihydrate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014465 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4465 1 g
11-101-4464 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4465 Supplier MilliporeSigma Supelco Supplier No. 118901G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym 4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate; Disodium bathophenanthrolinedisulfonate trihydrate

Specifications

CAS 52746-49-3
Linear Formula C24H14N2 Na2O6S2 · 3H2 O
Quantity 1 g
Formula Weight 590.53
Percent Purity ≥98.0% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.