Learn More
Cayman Chemical Bathophenanthroline, Purity: ≥98%, CAS: 1662-01-7, 10 g
SDP
Catalog No. 502911057
Encompass_Preferred
A colorimetric reagent for the detection of iron; can be used to quantify iron concentrations by colorimetric detection at 535 nm
Catalog No. 50-291-1057 Supplier Cayman Chemical Supplier No. 3481110 G
A colorimetric reagent for the detection of iron; can be used to quantify iron concentrations by colorimetric detection at 535 nm