Learn More
Medchemexpress LLC Batifiban TFA | 00-00-0 | 99.6% | 817.94 g/mol | C34H47N11O9S2 · xC2HF3O2 | 5 MG
SDP
Catalog No. 5000264433
Encompass_Preferred
Batifiban TFA is a cyclic peptide antagonist of platelet glycoprotein IIb/IIIa that inhibits platelet aggregation by blocking ligand binding such as vitronectin. Supplied as the trifluoroacetate (TFA) salt for research use, it is provided as a high-purity powder suitable for in vitro biochemical and cellular assays.
  • Antagonizes platelet glycoprotein IIb/IIIa to inhibit aggregation.
  • Blocks vitronectin binding to integrin receptors.
  • Supplied as trifluoroacetate (TFA) salt for stability.
  • High purity (~99.6%) suitable for biochemical assays.
  • Available in small research pack sizes, including 5 mg.
  • Store sealed, away from moisture and light; long-term storage at -80°C.
Catalog No. 50-002-64433 Supplier Medchemexpress LLC Supplier No. HY125053A5MG
Batifiban TFA is a cyclic peptide antagonist of platelet glycoprotein IIb/IIIa that inhibits platelet aggregation by blocking ligand binding such as vitronectin. Supplied as the trifluoroacetate (TFA) salt for research use, it is provided as a high-purity powder suitable for in vitro biochemical and cellular assays.
  • Antagonizes platelet glycoprotein IIb/IIIa to inhibit aggregation.
  • Blocks vitronectin binding to integrin receptors.
  • Supplied as trifluoroacetate (TFA) salt for stability.
  • High purity (~99.6%) suitable for biochemical assays.
  • Available in small research pack sizes, including 5 mg.
  • Store sealed, away from moisture and light; long-term storage at -80°C.