Learn More
Medchemexpress LLC HY-13564 5mg Medchemexpress, Batimastat CAS:130370-60-4 Purity:>98%
SDP
Catalog No. 502081394
Encompass_Preferred
Medchemexpress, HY-13564 5mg Batimastat CAS:130370-60-4 Batimastat is a potent broad spectrum MMP inhibitor with IC50 of 3, 4, 4, 6, and 20 nM for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1394 Supplier Medchemexpress LLC Supplier No. HY135645MG
Medchemexpress, HY-13564 5mg Batimastat CAS:130370-60-4 Batimastat is a potent broad spectrum MMP inhibitor with IC50 of 3, 4, 4, 6, and 20 nM for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.