Learn More
Selleck Chemical LLC Batimastat (BB-94) 10mg 130370-60-4
SDP
Catalog No. 501364465
Encompass_Preferred
Batimastat (BB-94) is a potent, broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3 with IC50 of 3 nM, 4 nM, 4 nM, 6 nM and 20 nM, respectively. Also inhibits the activitity of other metalloproteases, such as ADAM17. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4465 Supplier Selleck Chemical LLC Supplier No. S715510MG
Batimastat (BB-94) is a potent, broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-1, MMP-2, MMP-9, MMP-7 and MMP-3 with IC50 of 3 nM, 4 nM, 4 nM, 6 nM and 20 nM, respectively. Also inhibits the activitity of other metalloproteases, such as ADAM17. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.