Learn More
Apexbio Technology LLC Batimastat (BB-94)(Synonyms: BB-94, Batimastat, MMP inhibitor BB-94), 1mg, CAS: 130370-60-4.
SDP
Catalog No. 5000060882
Encompass_Preferred
Batimastat (BB-94 CAS 130370-60-4) is a synthetic small-molecule inhibitor targeting matrix metalloproteinases (MMPs) Structurally a polypeptide-like analogue of collagen substrates batimastat contains a peptidic backbone and a hydroxamate moiety that binds the catalytic zinc atom of MMPs It inhibits several MMP subtypes markedly including MMP-1 MMP-2 MMP-3 MMP-7 and MMP-9 with reported IC50 values of 3 4 20 6 and 4 nM respectively In preclinical studies batimastat demonstrates inhibitory effects on tumor growth and angiogenesis across various tumor models including ovarian and colon carcinoma xenografts making it relevant for cancer research and therapeutic development
Catalog No. 50-000-60882 Supplier Apexbio Technology LLC Supplier No. A25771
Batimastat (BB-94 CAS 130370-60-4) is a synthetic small-molecule inhibitor targeting matrix metalloproteinases (MMPs) Structurally a polypeptide-like analogue of collagen substrates batimastat contains a peptidic backbone and a hydroxamate moiety that binds the catalytic zinc atom of MMPs It inhibits several MMP subtypes markedly including MMP-1 MMP-2 MMP-3 MMP-7 and MMP-9 with reported IC50 values of 3 4 20 6 and 4 nM respectively In preclinical studies batimastat demonstrates inhibitory effects on tumor growth and angiogenesis across various tumor models including ovarian and colon carcinoma xenografts making it relevant for cancer research and therapeutic development