Learn More
Abcam Batimastat (BB-94), Matri x metalloprotease (MMP) inhibitor, 5MG
Catalog No. 502706342
Encompass_Preferred
MW 477.6 Da, Purity >98%. Potent, broad-spectrum MMP inhibitor (IC₅₀ values are 3 (MMP-1), 4 (MMP-2), 4 (MMP-9), 6 (MMP-7), 20 nM (MMP-3)). Anticancer activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6342 Supplier Abcam Supplier No. AB1420875MG
MW 477.6 Da, Purity >98%. Potent, broad-spectrum MMP inhibitor (IC₅₀ values are 3 (MMP-1), 4 (MMP-2), 4 (MMP-9), 6 (MMP-7), 20 nM (MMP-3)). Anticancer activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement