Learn More
Polysciences, Inc. Batsons #17 Anatomical Corrosion Kit
SDP
Catalog No. NC0225177
Encompass_Preferred
Low viscosity, acrylic-based resin system for room temperature preparation of anatomical corrosion castings. After hardening, tissue is corroded away by caustic solution, yielding a durable, scientifically exact model for anatomical study.
Catalog No. NC0225177 Supplier Polysciences, Inc. Supplier No. 073491
Low viscosity, acrylic-based resin system for room temperature preparation of anatomical corrosion castings. After hardening, tissue is corroded away by caustic solution, yielding a durable, scientifically exact model for anatomical study.