Learn More
MSC Clarke Floor Polisher, Ultra Speed, 20 in, Battery Powered, 2000 RPM
Catalog No. 502839297
Encompass
This floor polisher is designed for high-speed operation, featuring a 20-inch cleaning width and a battery-powered system for efficient maintenance. It functions as a walk-behind unit with pad assist drive, making it suitable for various floor care tasks.
  • Floor polisher for high-speed applications
  • Battery-powered operation
  • 20 in cleaning width
  • Durable polyethylene housing
  • 2.5 hp motor
  • 2000 RPM brush speed
  • Walk-behind operation with pad assist drive
Catalog No. 50-283-9297 Supplier MSC Supplier No. 35706449
Add to Cart
Add to Cart
Request a Quote
This floor polisher is designed for high-speed operation, featuring a 20-inch cleaning width and a battery-powered system for efficient maintenance. It functions as a walk-behind unit with pad assist drive, making it suitable for various floor care tasks.
  • Floor polisher for high-speed applications
  • Battery-powered operation
  • 20 in cleaning width
  • Durable polyethylene housing
  • 2.5 hp motor
  • 2000 RPM brush speed
  • Walk-behind operation with pad assist drive