Learn More
MSC Energizer Standard Battery, 9V, Alkaline, Miniature Snap Terminal, 6LR61
Catalog No. 502834873
Encompass
Energizer standard batteries are designed for reliable performance in a variety of devices. This 9V alkaline battery features a miniature snap terminal, making it suitable for applications requiring a dependable and long-lasting power source. It is disposable and adheres to IEC and ANSI standards.
  • Disposable battery for convenient use.
  • 9V alkaline chemistry for consistent power delivery.
  • Miniature snap terminal ensures secure connection.
  • Meets IEC and ANSI standards for quality and safety.
  • Industrial series for robust performance.
  • Rectangular shape for standard fitment.
  • 625 mAh capacity provides extended run time.
  • Operating temperature range of -18°C to 55°C for versatile use.
Catalog No. 50-283-4873 Supplier MSC Supplier No. 06505820
Energizer standard batteries are designed for reliable performance in a variety of devices. This 9V alkaline battery features a miniature snap terminal, making it suitable for applications requiring a dependable and long-lasting power source. It is disposable and adheres to IEC and ANSI standards.
  • Disposable battery for convenient use.
  • 9V alkaline chemistry for consistent power delivery.
  • Miniature snap terminal ensures secure connection.
  • Meets IEC and ANSI standards for quality and safety.
  • Industrial series for robust performance.
  • Rectangular shape for standard fitment.
  • 625 mAh capacity provides extended run time.
  • Operating temperature range of -18°C to 55°C for versatile use.